Learn More
Tocris Bioscience™ SU 3327
Selective JNK inhibitor
Brand: Tocris Bioscience™ 3607/50
809.19 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
SU 3327 is a selective inhibitor of c-Jun N-terminal kinase (JNK) (IC50 = 0.7 μM). Inhibits the protein-protein interaction between JNK and JIP (IC50 = 239 nM). Displays selectivity over p38 MAPK and Akt. Restores insulin sensitivity in a mouse model of type-2 diabetes. Displays antibiotic activity against a range of bacteria including S.aureus, A.baumanni, C.difficile and M.tuberculosis.Specifications
>99% | |
50 mg | |
261.3 |
Halicin | |
SU 3327 | |
SU 3327 |
For Research Use Only