missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SU 5402, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Potent and selective vascular endothelial growth factor receptor (VEGFR) and fibroblast growth factor receptor (FGFR) inhibitor (IC50 values are 0.02, 0.03, 0.51 and > 100 μM at VEGFR2, FGFR1, PDGFRβ and EGFR respectively). Inhibits embryonic left-right determination and exhibits potent anticancer activity in vitro and in vivo. Also attenuates integrin β4-induced differentiation of neural stem cells.
Specifications
Specifications
CAS | 215543-92-3 |
Molecular Formula | C17H16N2O3 |
Quantity | 1 mg |
Synonym | 3-3-2-carboxyethyl-4-methylpyrrol-2-methylidenyl-2-indolinone, z-3-4-methyl-2-2-oxoindolin-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, chembl89363, 3-3-2-carboxyethyl-4-methylpyrrol-2-yl methylene-2-indolinone, 3-4-methyl-2-3z-2-oxo-1h-indol-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, 3-4-methyl-2-2-oxo-1,2-dihydro-3h-indol-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, pubchem22440, kdr kinase inhibitor, 5, d0a4xj |
InChI Key | JNDVEAXZWJIOKB-JYRVWZFOSA-N |
SMILES | CC1=CNC(=C1CCC(=O)O)C=C2C3=CC=CC=C3NC2=O |
IUPAC Name | 3-[4-methyl-2-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid |
Molecular Weight (g/mol) | 296.326 |
PubChem CID | 5289418 |
Formula Weight | 296.32 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction