Learn More
Sulfur Standard Standard Metal Solution 10,000ppm, for ICP, Solution Aqueous, Fisher Chemical
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
MDL Number | 85316 |
Physical Form | Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
Product Identifier
- Sulfur standard solution 10 000 ppm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sulfur Standard Standard Metal Solution 10,000ppm, for ICP, Solution Aqueous, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction