Learn More
Hach Lange™ Sulphite Reagent Set
Brand: Hach Lange™ LCW054
This item is not returnable.
View return policy
Specifications
DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 | |
0.1 to 5 mg/L SO3 | |
Sulphite reagent set test |
50 | |
15°C to 25°C (59°F to 77°F) |