Learn More
Hach Lange™ Sulphite Reagent Set
Brand:  Hach Lange™ LCW054
Product Code. 10080754
  • 120.00 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
0.1 to 5 mg/L SO3
Sulphite reagent set test
50
15°C to 25°C (59°F to 77°F)