Learn More
Cytiva Superdex™ Peptide 3.2/300 Prepacked Precision Column
For high-resolution gel filtration of peptides and small biomolecules with molecular weights between 100 and 7000 in micropreparative scale
Brand: Cytiva 29036231
This item is not returnable.
View return policy
Description
- Prepacked with Superdex Peptide media
- Spherical composite of cross-linked agarose and dextran
- Supplied with two fingertight 1/16 in. male connectors for connection
- For sensitive and high resolution gel filtration of proteins, peptides, polynucleotides and other biomolecules
- Used for polishing step in purification process
- With new hardware design for direct connection to ÄKTATM systems
Specifications
0.01 to 0.15 mL/min. | |
13 to 15 μm | |
Superdex Pre-packed Column | |
Borosilicate Glass |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Superdex Peptide, 3.2/303
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol