Learn More
WTW™ Surfactants Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 252062
179.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
For waste water, seepage water, surface water | |
Certificates | |
0.05 to 1.50 mg/L | |
Photometric |
PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
25 Tests | |
c-Ten (cationic) surfactant cell test kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Surfactants Test Kits > Cell Test; c-Ten (Cationic); Detection Range: 0.05 to 1.50mg/L
Spot an opportunity for improvement?Share a Content Correction