Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Designed to accept SVL™ stirrer shafts with ground glass surfaces.
Brand:  SVL 588-12
 View more versions of this product
Product Code. 15557730
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Borosilicate glass complete with fittings
  • Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
  • Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SVL™ stirrer shaft
Quickfit™ ground Glass cone to allow connection to Quickfit™ flask
45/40
PTFE Based Polymer