missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talc, Pure, Fine Powder, Fisher Chemical™
50.90 € - 76.85 €
Chemical Identifiers
CAS | 14807-96-6 |
---|---|
Molecular Formula | H2Mg3O12Si4 |
Molecular Weight (g/mol) | 379.26 |
MDL Number | MFCD00792903 |
InChI Key | KNXVOGGZOFOROK-UHFFFAOYSA-N |
Synonym | magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate |
PubChem CID | 16211421 |
IUPAC Name | dioxosilane;oxomagnesium;hydrate |
SMILES | [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Description
Chemical Identifiers
14807-96-6 | |
379.26 | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
16211421 | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
dioxosilane;oxomagnesium;hydrate |
Specifications
800°C | |
White | |
H2Mg3O12Si4 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
379.26 | |
379.28g/mol |
14807-96-6 | |
Powder/Solid | |
MFCD00792903 | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
dioxosilane;oxomagnesium;hydrate | |
16211421 | |
Plastic powder jar |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title