Learn More
Tanshinone I, MedChemExpress
MedChemExpress Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).
Brand:  Medchem Express HY-N0134
Encompass_Preferred
 View more versions of this product
Product Code. 16468996
  • 376.65 € / 100mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).
Specifications
Specifications
Tanshinone I
Dark Brown
C18H12O3
Tanshinone A
O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC=C4C)=O
276.29
Research
COVID-19-immunoregulation
568-73-0
Research
100 mg
DMSO : 2 mg/mL (7.24 mM; Need ultrasonic)
276.29
98.0%
Solid