missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ hPSC Scorecard™ Kit, 384-well
TaqMan™ hPSC Scorecard™ Kit, 384-well
Brand: Applied Biosystems™ A15872
891.10 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
This item is not returnable.
View return policy
Description
The Applied Biosystems™ TaqMan™ hPSC Scorecard™ Kit 384w enables verification of trilineage differentiation potential for both human ES and iPS cell lines. The 384-well plate contains four sets of 94 predefined TaqMan™ Gene Expression assays (including endogenous controls) dried-down in the wells. Master mix is included for your convenience. To purchase the panel alone, see TaqMan™ hPSC Scorecard™ Panel 384w. This product is also available in a 2 x 96w Fast format.Assess an Entire Gene Signature in One Experiment
The TaqMan™ hPSC Scorecard™ Panel helps save time by offering pre-plated assays in a stable and convenient dried-down format. Simply add TaqMan™ Master Mix to your cDNA of interest and transfer to the 384-well plate with a multichannel pipette.
Compare Results to a Reference Standard
The TaqMan™ hPSC Scorecard™ Panel includes access to proprietary cloud-based analysis software that allows you to view and export graphs and tables of results, as well as generate reports. The software is compatible with seven Applied Biosystems™ qRT-PCR systems and is available at no additional charge.
Identify Lines with Trilineage Differentiation Potential
Include randomly differentiated embryoid body (EB) samples (differentiated for at least 7 days using standard EB methods) in your analysis and determine whether your lines are biased toward one or more of the three germ layers (endoderm, mesoderm, ectoderm).
Evaluate Inductive Differentiation Specified to One Germ Layer
The panel includes over twenty markers for each of the three germ layers that have been shown to respond as expected to directed differentiation in monolayer culture. Quickly evaluate time course data, culture conditions and media formulations for your germ layer of interest with this one panel.
Access Validated Content
The panel contents are based on published work (Bock et al., Cell 144, 439–452, 2011) and were validated against multiple human ES and iPS lines.
What You Get
The TaqMan™ hPSC Scorecard™ Panel 384w includes:
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
• One 384-well plate with four replicates of the 96-assay panel (room temperature) • One optical cover (room temperature) • One 5 mL vial of TaqMan™ Gene Expression Master Mix (2–8°C) |
|
Dried Down | |
Standard | |
TaqMan™ | |
Stem Cell Research | |
Stem Cell Analysis |
Primer-probe | |
384-well plate | |
QuantStudio™ 12k Flex, ViiA™ 7 System | |
1 x 384-well plate | |
Human | |
384 Reactions |
For Research Use Only. Not for use in diagnostic procedures.