Learn More
Cole-Parmer™ Essentials Cell Culturing Flask for BS-200 Series Cell Culture Stirring Systems
Product Code. 11553779
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 11553779
Brand: Cole-Parmer™ Essentials 2183730
Please to purchase this item. Need a web account? Register with us today!
This Cell Culturing Flask is a complete culture vessel—flask, stir rod, and caps—to use with biological stirrers.
  • Completely sealed vessel is ideal for pathogenic materials
Culture vessels come calibrated and have two side ports and a central filling port. All culture vessel parts can be autoclaved at 121°C and 15 psi for 30 minutes. Silicone-encapsulated stir rods reduce the possibility of cells attaching to and growing on the surface.
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
Height (English) 8.1 in.
Height (Metric) 205 mm
Autoclavable Autoclavable
Diameter (Metric) Outer 100 mm