Learn More
Medline™ Technico™ DIGIT-080 Sugar Pocket Refractometer
Brand: Medline™ 8100.0800M
This item is not returnable.
View return policy
Specifications
Pocket Sugar Refractometer | |
DIGIT-080 | |
1 Each |
0 to 80 Brix | |
Zero calibration with distilled water at 20° |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Medline™ Technico™ DIGIT-080 Sugar Pocket Refractometer > Brix Range: 0 to 80%
Spot an opportunity for improvement?Share a Content Correction