Learn More
Fisherbrand™ Temperature Probe
Monitor relative humidity and temperature with this probe. Fisherbrand™ Temperature Probes feature a polymer film sensor. For use with Fisherbrand hygrometers.
Brand: Fisherbrand™ 8779
This item is not returnable.
View return policy
Description
- Spare temperature probe
- For use with Hygrometer FB70257
- Length: 21.5cm
- Dia. 1.67cm
Order Info
dimForUseWithEquipment
Specifications
Replacement Temperature Probe | |
FB70257 | |
1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Temperature Probe > For use with: FB70257; Quantity: 1Each
Spot an opportunity for improvement?Share a Content Correction