Learn More
Memmert™ Temperature Restriction for UN260plus
Optional temperature restriction between 60°C and 250°C.
Brand:  Memmert™ UN260plusA8
Product Code. 17549995
  • 164.00 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Accessory
  • 1 Choice
  • Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
Specifications
UN260plus Oven
Temperature Restriction
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Temperature Restriction for UN260plus > For Use With: UN260plus