Learn More
Memmert™ Temperature Restriction for UN260plus
Optional temperature restriction between 60°C and 250°C.
Brand: Memmert™ UN260plusA8
This item is not returnable.
View return policy
Description
- Accessory
- 1 Choice
- Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
UN260plus Oven | |
Temperature Restriction |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Temperature Restriction for UN260plus > For Use With: UN260plus
Spot an opportunity for improvement?Share a Content Correction