missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TER-119 Rat, Qfluor 710, Clone: TER-119, enQuireBio™
Rat Monoclonal Antibody
124.00 € - 263.00 €
Specifications
Antigen | TER-119 |
---|---|
Clone | TER-119 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962448
|
EnquireBio
QAB76-QF710-25ug |
25 μg |
124.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15952448
|
EnquireBio
QAB76-QF710-100ug |
100 μg |
263.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
TER-119 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ly76 | |
Primary | |
Store at 2-4°C. |
TER-119 | |
Flow Cytometry | |
Qfluor 710 | |
RUO | |
104231 | |
IgG2b κ | |
Protein A/G Purified |