Learn More
TES-1025, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
Specifications
Specifications
| Chemical Name or Material | TES-1025 |
| CAS | 1883602-21-8 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H13N3O3S2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (260.80 mM; Need warming) |
| SMILES | O=C1C(C#N)=C(C2=CC=CS2)N=C(SCC3=CC=CC(CC(O)=O)=C3)N1 |
| Molecular Weight (g/mol) | 383.44 |
| Formula Weight | 383.44 |
| Show More |