missing translation for 'onlineSavingsMsg'
Learn More

Tesa™ Fabric Tape

POA

Specifications

Product Type Tesa Fabric Tape
Dimensions (LxW) 50 m x 19 mm
Length (Metric) 50 m
Width (Metric) 19 mm
View More Specs

Products 3
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10087741
Encompass
View Documents
TESA
TESA57230BLAU
Blue
POA
1 roll
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

10129451
Encompass
View Documents
TESA
TESA57230ROT
Red
POA
1 roll
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

10353272
Encompass
View Documents
TESA
TESA57230GELB
Yellow
POA
1 roll
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Specifications

Specifications

Tesa Fabric Tape
50 m x 19 mm
50 m
19 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.