Learn More
Hach™ Test 'N Tube (TNT) Cuvette Tests
Product Code. 10634535
Click to view available options
For Use With (Application):
COD Determination
Nitrite Determination
Unit Size:
Each
Pack of 25
This item is not returnable. View return policy
Product Code. 10634535
Brand: Hach™ 2608345
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Provides a self-contained package for mixing and measurement. Hach™ Test 'N Tube (TNT) Hach™ Test 'N Tube (TNT) Cuvette Tests include 16mm capped tubes and all necessary reagents in one convenient package for reliable results and easy handling.
Provides a self-contained package for mixing and measurement. Hach™ Test 'N Tube (TNT) Hach™ Test 'N Tube (TNT) Cuvette Tests include 16mm capped tubes and all necessary reagents in one convenient package for reliable results and easy handling.
Test 'N Tube products enable convenient testing with ready to use ampules and pre-measured reagents all packed into one convenient package. Available for determinations of a variety of parameters.
  • Test 'N Tube: Capped 16mm vials provide a safe, self-contained package for mixing and measurement
  • Powder Pillows: Premeasured, single dose packets of powdered reagent
  • Dropper bottles: For easy addition of liquid reagents
  • Precisely dosed reagents and test cuvettes for easy planning and inventory control
Specifications
Description Colorimetric determination cuvette tests
For Use With (Application) Nitrite Determination
For Use With (Equipment) Hach DR900
Includes 16mm Tubes and all necessary reagents
No. of Tests 50
Quantity 1 Kit
Range 0.003 to 0.500mg/L NO2-N
Test Method NitriVer 3 Diazotization
Type Test 'N Tube