Learn More
Carl Roth™ Test sticks
Brand:  Carl Roth™ L829.1
Encompass
Product Code. 10312452
  • 38.85 € / Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
H2O2
0.2 mm-thick plastic strip with testing paper affixed to the lower end.
Single-use
Coloration on test paper is compared to a color scale on the side of the can
Test Sticks
0.5 to 25 mg/L H2O2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Carl Roth™ Test sticks