Learn More
Grant Instruments™ Test Tube Tray for Grant Shaking Water Baths
Click to view available options
Type:
Flask Tray
Test Tube Tray
Test Tube/Microtube Tray
For Use With (Equipment):
LSB12
LSB18
OLS26
Unit Size:
Each
Descrizione
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruemnts™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
Test Tube Tray for Grant Shaking Water Baths
- TS12: Holds up to 3 SR Racks for LSB12 Model Aqua Pro Shaking Water Bath
- TS18: Holds up to 5 SR Racks for LSB18 Model Aqua Pro Shaking Water Bath
- TS26: Holds up to 5 SR Racks for OLS26 Model Aqua Pro Shaking Water Bath
Specifica
Specifica
| For Use With (Equipment) | LSB12 |
| Type | Test Tube Tray |
| Holds | Up to 5 SR Racks |
| Material | Stainless Steel |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto