Learn More
TET3 Mouse anti-Human, Clone: OTI4B7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803982

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TET3, a member of the ten-eleven-translocation (TET) family of genes, is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine and is most abundantly expressed in hematopoietic cells. Unlike the related TET2 protein, mutations in TET3 have not been observed in any myeloid maligcies. TET3 has been shown to be involved in the demethylation of zygotic DNA before the first mitosis and has been suggested to be involved in the epigenetic reprogramming of the zygotic paternal DNA following natural fertilization and may also contribute to somatic cell nuclear reprogramming during animal cloning.Specifications
TET3 | |
Monoclonal | |
Unconjugated | |
TET3 | |
hCG_40738 | |
Mouse | |
Affinity Chromatography | |
RUO | |
200424 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43151 | |
TET3 | |
Human recombit protein fragment corresponding to amino acids 241-568 of human TET3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.