Learn More
Tetrahydrothiopyran-4-carbonyl chloride, 95%, Thermo Scientific™
Product Code. 10452314
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. 10452314
Brand: Thermo Scientific Maybridge CC61902DA
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
Chemical Identifiers
CAS 121654-84-0
Molecular Formula C6H9ClOS
Molecular Weight (g/mol) 164.647
MDL Number MFCD09879922
InChI Key RJRIIGFPFQOTNF-UHFFFAOYSA-N
Synonym tetrahydrothiopyran-4-carbonyl chloride, tetrahydro-thiopyran-4-carbonyl chloride, tetrahydro-2h-thiopyran-4-carbonyl chloride, 2h-thiopyran-4-carbonylchloride, tetrahydro, acmc-20mplw
PubChem CID 14547368
IUPAC Name thiane-4-carbonyl chloride
SMILES C1CSCCC1C(=O)Cl
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 164.66
Percent Purity 95%
Chemical Name or Material Tetrahydrothiopyran-4-carbonyl chloride