missing translation for 'onlineSavingsMsg'
Learn More
Tetrakis(dimethylamino)silane, 99%, Thermo Scientific Chemicals Product Code.: 11486160

Tetrakis(dimethylamino)silane, 99%, Thermo Scientific Chemicals

Product Code. 11486160
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11486160

Brand: Thermo Scientific Alfa Aesar H27092.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1624-01-7
Molecular Formula C8H24N4Si
Molecular Weight (g/mol) 204.39
MDL Number MFCD00014859
InChI Key SSCVMVQLICADPI-UHFFFAOYSA-N
Synonym tetrakis dimethylamino silane, silanetetramine, octamethyl, octamethylsilanetetramine, octamethylsilatetramine, silane, tetrakis dimethylamino, silane 48-12 tetrakis, dimethylamine, n,n',n,n'-silanetetrayltetrakis, silanetetramine, n,n,n',n',n,n,n',n'-octamethyl, n,n,n',n',n,n,n',n'-octamethylsilanetetraamine, si nme2 4
PubChem CID 74194
IUPAC Name N-methyl-N-[tris(dimethylamino)silyl]methanamine
SMILES CN(C)[Si](N(C)C)(N(C)C)N(C)C

Specifications

Boiling Point 196°C
Melting Point 15°C
Quantity 1 g
Linear Formula [(CH3)2N]4Si
UN Number UN3129
Flash Point 58°C (136°F)
Solubility Information Reacts with water.
Formula Weight 204.4
Percent Purity 99%
Odor Amine-like
Refractive Index 1.443
Sensitivity Moisture sensitive
Density 0.885
Chemical Name or Material Tetrakis(dimethylamino)silane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10181863 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27092.03,H27092.09
Lot Number10166820 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27092.03,H27092.09
Lot Number10224757 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27092.03,H27092.09
Lot Number10192049 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27092.03,H27092.09
Lot Number10175216 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27092.03,H27092.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrakis(dimethylamino)silane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.