Learn More
Thermo Scientific TG-BOND Alumina Na2SO4 PLOT Column 25m x 0.32mm x5um
Brand:  Thermo Scientific 26001-5970
TG-BOND Alumina Na2SO4 PLOT Column 25m x 0.32mm x5um
Product Code. 15890890
  • ONLINE EXCLUSIVE PRICE
    1025.00 €
    1019.88 € / Each
    Save 5.12 €
    0.50% Off
Only null left
Estimated Shipment: 03-02-2026