Learn More
Abnova™ TGFBR3 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FS0051.100uL
This item is not returnable.
View return policy
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
BGCAN,betaglycan | |
7049 | |
100 μL |
Tube | |
transforming growth factor, beta receptor III | |
TGFBR3 | |
DNA/RNA | |
Liquid |