Learn More
Epredia™ Richard-Allan Scientific™ Cyto-Stain™ G
Product Code. 12628106 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 12628106
Brand: Epredia™ 7501G
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Produces results similar to those achieved with conventional Papanicolaou stains
  • Can be used for all gynecological and non-gynecological procedures
  • Designed to produce greener cyanophilic hues in intermediate and basal cells
  • Requires one counterstain instead of two
  • Reduces staining time
TRUSTED_SUSTAINABILITY
Specifications
Type Cytostain
Quantity 1 gal.