Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Documention
Shop All Thermo Scientific ProductsDescripción
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Especificaciones
Especificaciones
| For Use With (Equipment) | Thermo Scientific Ultra-Low Temperature Freezers including all Thermo Scientific Ultra-Low Temperature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE |
| Description | Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | IQ/OQ Documentation |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido