Learn More
Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces
Product Code. p-6024369
Click to view available options
Capacity (Metric):
5 L
Unit Size:
Each
Product Code. 10509722
Brand: Thermo Scientific™ F4801033
Only null left
Add to basket
Add to basket
Ideal for general laboratory use, including gravimetric analysis, sintering, quantitative analysis and heat treating.
Ideal for general laboratory use, including gravimetric analysis, sintering, quantitative analysis and heat treating.
  • Reaches a 1200°C maximum temperature
  • Built-in vent port removes contaminants and moisture to extend the life of the heating element and furnace
  • Ideal for ashing applications • For added protection, the door safety switch stops power to heating elements when door opens
  • Thermocouple break protection cuts power to heating elements, preventing a thermocouple failure runaway condition
  • Two open coil heating elements on chamber sides assure fast heat-up with minimum temperature gradient Thermal-efficient ceramic insulation surrounds chamber for maximum energy efficiency
  • 0.312 in. dia. port for monitoring chamber temperatures with independent measuring device at rear of chamber 
TRUSTED_SUSTAINABILITY
Specifications
Controller Type A1
Capacity (Metric) 5 L
Temperature (Metric) 100°C to 1200°C
Plug Type EU plug: CEE 7/7
Dimensions (D x W x H) Interior 25 x 18 x 13 cm (10 x 7 x 5 in.)
Dimensions (L x W x H) Exterior 50 x 34 x 19 cm (19.5 x 13.3 x 19 in.)
Electrical Requirements 240 V 50/60 Hz
Wattage 1560 w
Amperage 6.5 A