Learn More
Karl Hecht™ Thermometer, Red Petrol-Filled
General purpose, normal glass, enclosed scale thermometer. Karl Hecht™ Thermometer, Red Petrol-Filled is easily readable.
Brand: Karl Hecht™ 43201040
This item is not returnable.
View return policy
Description
- Red Petrol-filled for easy readability.
Specifications
Normal Glass | |
Red Petrol | |
Dry Thermometer 250 | |
250°C |