Learn More
Sonoco™ ThermoSafe PolarPack™ Foam Bricks
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
Brand:  Sonoco™ FPP20
 View more versions of this product
Product Code. 17879271
Description
Description
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
  • Provide refrigeration for transport of food, pharmaceuticals, and medical products.
  • Gel will phase change at 0° C.
  • PolarPack Foam Bricks maintain designated packing configurations throughout the trip.
  • For frozen or refrigerated shipments.
  • Alternative to wet ice.
  • Available in several sizes
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    32/Cs.
    17 oz.
    9 x 4.75 x 0.75 in. (22.8 x 12.1 x 1.9 cm)
    0°C
    PolarPack Foam Brick
    482 g
    Semi-rigid Foam
    Non-mechanical refrigeration
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Sonoco™ ThermoSafe PolarPack™ Foam Bricks > 17 oz.