Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key threaded, SBW -J glass
Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.4 NS
18.8 NS
Unit Size:
Pack of 100
Description
The 3-part tap safety device holds and secures the threaded tap plug in the tap sleeve.
- 3-part, complete for SBW-J tap plug
- Greater safety through installation of the 3-part screw locking device
Specifications
Specifications
| Material | Polypropylene |
| Autoclavable | Not Autoclavable |
| Closure Size | 18.8 NS |
| No. per Pack | 100/Pk. |
| For Use With (Application) | For securing the threaded tap plug in the ground DURAN™ one-way tap |