missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Thromboxane B2 Parameter Assay Kit

Product Code. 18712424
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18712424

Brand: R&D Systems™ KGE011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thromboxane B2 Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Thromboxane B2 Parameter Assay Kit quantifies multi-species Thromboxane B2 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.31 - 20ng/mL
Assay Sensitivity 0.31ng/mL
Conjugate Biotin
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity TXB2 Levels in Various Samples
For Use With (Application) ELISA
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.