missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TIRAP (TLR2 and TLR4) Inhibitor Peptide Set

Product Code. 18146244 Shop All Bio Techne Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18146244

Brand: Novus Biologicals™ NBP2293315MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in research applications

TRUSTED_SUSTAINABILITY

Specifications

Host Species Human, Mouse
Components TIRAP Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKLQLRDAAPGGAIVS (TIRAP sequence is underlined). Molecular weight: 3701.4. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361.
For Use With (Application) Inhibition of TIRAP binding to TLR2 or TLR4
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
Molecular Weight (g/mol) 3701.4
Inhibitors TLR2, TLR4
Form Lyophilized

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.