Learn More
Sakura Finetek™ Tissue-Tek™ Cryomold™ Biopsy Molds
Disposable plastic Cryomold™ molds create a uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound for frozen sectioning.
Brand:  Sakura Finetek™ 4565
Encompass
Product Code. 10690461
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Biopsy mold requires an adapter for the Cryobar™ freezing wells of the Tissue Tek II™
  • 10 x 10 x 5mm
  • 100/pack
  • Combines excellent quality with economical pricing
Specifications
Specifications
Biopsy Mold
10 x 10 x 5 mm
Frozen sectioning, specimen blocks
Plastic
Single-use
100/Pk.