Learn More
Abnova™ TNFRSF13C 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Brand:  Abnova™ H00115650-T01.100uL
Product Code. 16167233
  • 215.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Specifications
293T
TNFRSF13C 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 20.35kDa
Transient overexpression cell lysate was tested with anti-TNFRSF13C antibody (Mfr. No. H00115650-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_052945.2
pCMV-TNFRSF13C full-length