Learn More
Tocris Bioscience™ Amylin
Endogenous peptide agonist for amylin, calcitonin, CGRP and adrenomedullin receptors
Brand:  Tocris Bioscience™ 3418/500U
Product Code. 18055372
  • 367.00 € / 500µg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Amylin is an endogenous peptide agonist for amylin, calcitonin, CGRP and adrenomedullin receptors. Inhibits glucagon secretion, delays gastric emptying and acts as a satiety agent. Displays glucose lowering effects in vivo.
Specifications
Specifications
500 μg