missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.7% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Specifications
Specifications
Density | 0.865g/mL at 25°C |
Quantity | 25 L |
Explosion Limit | 0.07 |
Flash Point | 4°C (39.2°F) |
Ignition Point | 997°F |
Linear Formula | C6H5CH3 |
Vapor Pressure | 22mmHg (20°C), 26mmHg (25°C) |
UN Number | 1294 |
Beilstein | 635760 |
Formula Weight | 92.14g/mol |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Safety and Handling



Product Identifier
- Toluene
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Toluene, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.7% (GC), Honeywell Riedel-de Haën™ >
Spot an opportunity for improvement?Share a Content Correction