missing translation for 'onlineSavingsMsg'
Learn More

Tosufloxacin tosylate hydrate, MedChemExpress

Product Code. 16432359
Encompass_Preferred
Click to view available options
Quantity:
200 mg
1 g
5 g
10 g
Unit Size:
10g
1g
200mg
5g
This item is not returnable. View return policy

Product Code. 16432359

Brand: Medchem Express HYB1802A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tosufloxacin (tosylate hydrate) is a fluoroquinolone antibacterial agent.

Tosufloxacin (tosylate hydrate) is a fluoroquinolone antibacterial agent.

Specifications

Chemical Name or Material Tosufloxacin tosylate hydrate
Physical Form Solid
CAS 1400591-39-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C26H25F3N4O7S
For Use With (Application) COVID-19-immunoregulation
Synonym A-61827 tosylate hydrate
Solubility Information DMSO : 12.5 mg/mL (21.02 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(S(=O)(O)=O)C=C1.O=C(C2=CN(C3=CC=C(F)C=C3F)C4=C(C=C(F)C(N5CC(N)CC5)=N4)C2=O)O.O
Molecular Weight (g/mol) 594.56
Formula Weight 594.56
Percent Purity 99.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.