Learn More
Bellingham and Stanley™ Touch Screen Refractometer
Click to view available options
Description:
Resolution: 0.01 Brix
Resolution: 0.01/0.001 Brix
Unit Size:
Each
Description
Touch Screen Refractometer with Peltier temperature control features the latest integral 7 in. full color touchscreen, high definition display, and USB/LAN connectivity. The refractometer is supplied with two RFID tags, instruction manual, and certificate of conformity. Perfect for measurement and control in demanding food, beverage, chemical and industrial applications.
- Scales
- RI 1.32 - 1.58
- Brix 0 - 100
- Resolution:
- RI 0.00001 (RFM330-T)/ 0.000001 (RFM340-T)
- Brix 0.01
Specifications
Specifications
| Type | Digital Refractometer |
| Scale Range | 0 to 100 Brix |
| Description | Resolution: 0.01/0.001 Brix |
| Model | RFM340-T |
| Accuracy | ± 0.01 to 0.03 Brix |
| Includes | Refractometer, RFID Tags (2), Instruction Manual, and Certificate of Conformity |