missing translation for 'onlineSavingsMsg'
Learn More

TPPB, Tocris Bioscience™

Product Code. 30000864
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity PKC activator; also APP modulator

High affinity protein kinase C activator (Ki =11.9 nM). Induces differentiation of hESCs into Pdx-1 expressing pancreatic progenitor cells. Also enhances secretion of amyloid precursor protein. Cell permeable.

Specifications

CAS 497259-23-1
Molecular Formula C27H30F3N3O3
Linear Formula C27H30F3N3O3
Quantity 1 mg
Synonym tppb, alpha-app modulator, 2s,5s-e,e-8-5-4-trifluoromethyl phenyl-2,4-pentadienoylamino benzolactam, 2e,4e-n-2s,5s-1,2,3,4,5,6-hexahydro-5-hydroxymethyl-1-methyl-2-1-methylethyl-3-oxo-1,4-benzodiazocin-8-yl-5-4-trifluoromethyl phenyl-2,4-pentadienamide, 2e,4e-n-2s,5s-5-hydroxymethyl-2-isopropyl-1-methyl-3-oxo-2,4,5,6-tetrahydro-1,4-benzodiazocin-8-yl-5-4-trifluoromethyl phenyl penta-2,4-dienamide, 5-4-trifluoromethyl-phenyl-penta-2,4-dienoic acid 5-hydroxymethyl-2-isopropyl-1-methyl-3-oxo-1,2,3,4,5,6-hexahydro-benzo e 1,4 diazocin-8-yl-amide
Solubility Information Soluble to 50mM in DMSO
InChI Key WOLVEMPZUIFSII-IHHOKICGSA-N
SMILES CC(C)C1C(=O)NC(CC2=C(N1C)C=CC(=C2)NC(=O)C=CC=CC3=CC=C(C=C3)C(F)(F)F)CO
IUPAC Name (2E,4E)-N-[(2S,5S)-5-(hydroxymethyl)-1-methyl-3-oxo-2-propan-2-yl-2,4,5,6-tetrahydro-1,4-benzodiazocin-8-yl]-5-[4-(trifluoromethyl)phenyl]penta-2,4-dienamide
Molecular Weight (g/mol) 501.55
PubChem CID 9935767
Formula Weight 501.54
Physical Form Solid
Chemical Name or Material (2E,4E)-N-[(2S,5S)-1,2,3,4,5,6-Hexahydro-5-(hydroxymethyl)-1-methyl-2-(1-methylethyl)-3-oxo-1,4-benz
Grade >98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.