Learn More
Thermo Scientific™ TR-FRET Coregulator Buffer A
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Brand: Thermo Scientific™ PV4384
This item is not returnable.
View return policy
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Specifications
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays | |
TR-FRET Coregulator Buffer A may be shipped frozen or at room temperature. Store at room temperature (20°C to 30°C). | |
Liquid |
TR-FRET Coregulator Buffer A, 25mL, Liquid, 7.5 pH, 20 to 30°C Storage, Room Temperature or on Wet or Dry Ice Shipping Conditions, Water Soluble, For use with Pharma and Biopharma, Target and Lead Identification and Validation | |
25 mL | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3121336C | Certificate of Analysis | 06/03/2025 | PV4384 |
3121336B | Certificate of Analysis | 04/03/2025 | PV4384 |
3028856B | Certificate of Analysis | 21/02/2025 | PV4384 |
3121336A | Certificate of Analysis | 21/02/2025 | PV4384 |
2718987B | Certificate of Analysis | 14/09/2023 | PV4384 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ TR-FRET Coregulator Buffer A > 25 mL