Learn More
Thermo Scientific™ TR-FRET Coregulator Buffer B
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Brand:  Thermo Scientific™ PV4418
Product Code. 10418853
  • 154.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays
Pharma and biopharma, target and lead identification and validation, drug discovery and development, nuclear receptor biology, biochemical nuclear receptor assays, nuclear receptor coregulator interaction
TR-FRET Coregulator Buffer B may be shipped frozen or at room temperature.Store at room temperature (20°C to 30°C).
Liquid
Water soluble
TR-FRET Coregulator Buffer B, 25mL, Liquid, Store at 20 to 30°C, Water Soluble, Used For Pharma & Biopharma, Target & Lead Identification & Validation
25 mL
Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2217449C Certificate TypeCertificate of Analysis Date28/05/2020 Catalog NumberPV4418
Lot Number1995248B Certificate TypeCertificate of Analysis Date14/09/2019 Catalog NumberPV4418
Lot Number1995248A Certificate TypeCertificate of Analysis Date30/07/2019 Catalog NumberPV4418
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ TR-FRET Coregulator Buffer B > 25 mL