Learn More
Thermo Scientific™ TR-FRET Coregulator Buffer D
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Brand: Thermo Scientific™ PV4420
This item is not returnable.
View return policy
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Specifications
TR-FRET Coregulator Buffer D, 25mL, Liquid, 7.5 pH, 4°C Storage, Room Temperature or on Wet or Dry Ice Shipping Conditions, Water Soluble, For use with Protein Assays and Analysis, High Throughput-Compatible, Enzyme Assay | |
Liquid | |
Water | |
7.5 | |
TR-FRET Coregulator Buffer D may be shipped frozen or at room temperature. Store at 4°C. |
Pharma and biopharma, target and lead identification and validation, Lanthascreen™ nuclear receptor cofactor interaction assays | |
25mL, 25 mL | |
Nuclear Receptor Buffer | |
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3149897B | Certificate of Analysis | 02/04/2025 | PV4420 |
3149897A | Certificate of Analysis | 11/03/2025 | PV4420 |
2258748E | Certificate of Analysis | 04/04/2024 | PV4420 |
2258748D | Certificate of Analysis | 23/02/2024 | PV4420 |
2258748C | Certificate of Analysis | 01/02/2024 | PV4420 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ TR-FRET Coregulator Buffer D > 25 mL