missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Amine Receptor 1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93938-0.1ml
This item is not returnable.
View return policy
Beschreibung
Trace Amine Receptor 1 Polyclonal antibody specifically detects Trace Amine Receptor 1 in Human, Mouse, Rat samples. It is validated for Western Blot
Spezifikation
| Trace Amine Receptor 1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAAR1 (NP_612200.1). TSTDIMLSSASIFHLSFISIDRYYAVCDPLRYKAKMNILVICVMIFISWSVPAVFAFGMIFLELNFKGAEEIYYKHVHCRGGCSVFFSKISGVLTFMTSFY | |
| 0.1 mL | |
| GPCR | |
| 134864 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur