Learn More
trans-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
218.09 € - 616.07 €
Chemical Identifiers
CAS | 1212272-03-1 |
---|---|
Molecular Formula | C13H15NO4 |
Molecular Weight (g/mol) | 249.27 |
MDL Number | MFCD03844619 |
InChI Key | ZNRUEEQIUVKKBL-UHFFFAOYNA-N |
Synonym | trans-2-benzyloxycarbonylaminocyclobutanecarboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, trans-2-cbz-amino cyclobutane-1-carboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane carboxylic acid, 1r,2r-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid |
PubChem CID | 24720935 |
IUPAC Name | (1R,2R)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid |
SMILES | OC(=O)C1CCC1NC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468006
|
Thermo Scientific Alfa Aesar
H62919.MD |
250 mg |
218.09 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15458006
|
Thermo Scientific Alfa Aesar
H62919.03 |
1 g |
616.07 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesIncompatible with oxidizing agents.
Chemical Identifiers
1212272-03-1 | |
249.27 | |
ZNRUEEQIUVKKBL-UHFFFAOYNA-N | |
24720935 | |
OC(=O)C1CCC1NC(=O)OCC1=CC=CC=C1 |
C13H15NO4 | |
MFCD03844619 | |
trans-2-benzyloxycarbonylaminocyclobutanecarboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, trans-2-cbz-amino cyclobutane-1-carboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane carboxylic acid, 1r,2r-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid | |
(1R,2R)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid |
Specifications
1212272-03-1 | |
MFCD03844619 | |
ZNRUEEQIUVKKBL-UHFFFAOYNA-N | |
(1R,2R)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid | |
24720935 | |
97% |
C13H15NO4 | |
trans-2-benzyloxycarbonylaminocyclobutanecarboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, trans-2-cbz-amino cyclobutane-1-carboxylic acid, trans-2-benzyloxycarbonylaminocyclobutane carboxylic acid, 1r,2r-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid | |
OC(=O)C1CCC1NC(=O)OCC1=CC=CC=C1 | |
249.27 | |
249.26 | |
trans-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.