missing translation for 'onlineSavingsMsg'
Learn More
trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%, Thermo Scientific Chemicals Product Code.: 15447446

trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%, Thermo Scientific Chemicals

Product Code. 15447446
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15447446

Brand: Thermo Scientific Alfa Aesar H62444.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149423-77-8
Molecular Formula C14H20N2O2
Molecular Weight (g/mol) 248.33
MDL Number MFCD03839901,MFCD06657668,MFCD07772066
InChI Key JQVBZZUMWRXDSQ-UHFFFAOYSA-N
Synonym benzyl cis-4-aminocyclohexyl carbamate, benzyl trans-4-aminocyclohexyl carbamate, n-cbz-trans-1,4-cyclohexanediamine, cis-1-n-cbz-1,4-cyclohexyldiamine, cis-n-cbz-1,4-cyclohexanediamine, 4-amino-cyclohexyl-carbamic acid benzyl ester, benzyl cis-4-aminocyclohexylcarbamate, benzyl 4-aminocyclohexyl carbamate, benzyl 1r,4r-4-aminocyclohexyl carbamate, 1-n-cbz-trans-1,4-cyclohexyldiamine
PubChem CID 18650082
IUPAC Name benzyl N-(4-aminocyclohexyl)carbamate
SMILES NC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 248.32
Percent Purity 97%
Chemical Name or Material trans-4-(Benzyloxycarbonylamino)cyclohexylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.