missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Transparent Natural Rubber Latex Tubing
Natural Rubber Latex Tubing that is sterilizable at 120°C.
Brand: Deutsch Neumann™ 3050407
This item is not returnable.
View return policy
Description
- Hardness of 40 Shore A
- Light and flexible
- Offers excellent tensile strength, elongation, resilience and tear resistance
- Available in 10 m rolls
Specifications
ISO 9001:2008 | |
Natural Rubber | |
10 m | |
Non-sterile |
4 mm | |
7 mm | |
1.5 mm | |
Transparent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction