missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetone
Methyl Ethyl Ketone
Unit Size:
Each
Pack of 12
Pack of 6
Description
Working with chemical substances that can be dangerous requires a high level of responsibility and care. VITsafe™ safety wash-bottles fulfill all safety requirements for safe handling. Spray bottles are individually wrapped in clearly labeled polyethylene bags.
Constructed of durable, transparent polypropylene
Constructed of durable, transparent polypropylene
- PP adds a significantly higher level of resistance to many solvents, including acetone and MEK
- Wash bottles and spray tubes made from PP may be autoclaved at 121°C for 20 minutes
- Transparent material allows for easy identification and monitoring
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- In accordance to EC Directive No. 1272/2008 (GHS)
- Design eliminates almost all static overpressure
- Prevents virtually all leaking or dripping that can occur with temperature changes in the lab
- Color-coded caps support easy visual identification of the media
- Bottles utilize screw cap lids with GL threads for a strong, secure hold
- Finely drawn tip creates a precise spray jet for fine control
- Optimized medium backflow through the continuous spray tube
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Transparent |
| Material | Polypropylene |
| Packaging | Individually wrapped in PE bag |
| Quantity | 12Pack |
| Screw Color | Red |
| Thread Size (Metric) | GL 25 |
| Mouth | Narrow |
| Height (Metric) | 135 mm |
| Product Type | Safety Wash Bottle |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction