missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles

Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP and precise spray control to eliminate drips and leaks. Constructed of chemically resistant material for use with acetone and MEK.

Brand:  VITLAB™ 1431829

 View more versions of this product

Product Code. 11817900

  • 104.00 € / Pack of 12
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Working with chemical substances that can be dangerous requires a high level of responsibility and care. VITsafe™ safety wash-bottles fulfill all safety requirements for safe handling. Spray bottles are individually wrapped in clearly labeled polyethylene bags.

Constructed of durable, transparent polypropylene
  • PP adds a significantly higher level of resistance to many solvents, including acetone and MEK
  • Wash bottles and spray tubes made from PP may be autoclaved at 121°C for 20 minutes
  • Transparent material allows for easy identification and monitoring
Permanent safety imprint on the bottle with all important information
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No, hazard pictogram, signal word
  • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
  • In accordance to EC Directive No. 1272/2008 (GHS)
Metal-free, color-colded VENT-CAP
  • Design eliminates almost all static overpressure
  • Prevents virtually all leaking or dripping that can occur with temperature changes in the lab
  • Color-coded caps support easy visual identification of the media
  • Bottles utilize screw cap lids with GL threads for a strong, secure hold
Optimized for precise spray control
  • Finely drawn tip creates a precise spray jet for fine control
  • Optimized medium backflow through the continuous spray tube
Specifications

Specifications

250 mL
Polypropylene
12Pack
GL 25
135 mm
Acetone
Vented Screw Cap
Transparent
Individually wrapped in PE bag
Red
Narrow
Safety Wash Bottle
58 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles > Chemical Label: Acetone, Capacity: 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.