Learn More
VITLAB™ Transparent PTFE Watch Glasses
Brand:  VITLAB™ 113297
Product Code. 10294501
  • 24.50 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Autoclavable
Round
Used as Beaker Covers
PTFE
Watch Glass
75 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VITLAB™ Transparent PTFE Watch Glasses > Diameter: 75mm; White